COM448613806 Combi-Blocks, Inc.

3-Acetoxymethylphenylboronic acid, pinacol ester

  • MDL number MFCD02179491
  • Cas Number : 562098-07-1
  • Molecular Weight : 276.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2485

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2485
Availability details for
Quantity
  • Availability
  • Add to Cart