COM448613810 Combi-Blocks, Inc.

2-(2-Bromoethyl)phenylboronic acid

  • MDL number MFCD06659854
  • Cas Number : 850568-82-0
  • Molecular Weight : 228.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2489

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2489
Availability details for
Quantity
  • Availability
  • Add to Cart