COM448613813 Combi-Blocks, Inc.

4-(2-Bromoethyl)phenylboronic acid

  • MDL number MFCD12546453
  • Cas Number : 137756-90-2
  • Molecular Weight : 228.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2493

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2493
Availability details for
Quantity
  • Availability
  • Add to Cart