COM448613828 Combi-Blocks, Inc.

4-Borono-3-cyclohexen-1-one, ethylene glycol ketal

  • MDL number MFCD06659855
  • Cas Number : 850567-90-7
  • Molecular Weight : 184.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2533

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2533
Availability details for
Quantity
  • Availability
  • Add to Cart