COM448613836 Combi-Blocks, Inc.

2-Chloro-3-propoxyphenylboronic acid

  • MDL number MFCD16660271
  • Cas Number : 1256345-51-3
  • Molecular Weight : 214.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2556

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2556
Availability details for
Quantity
  • Availability
  • Add to Cart