COM448613838 Combi-Blocks, Inc.

2-Butylthio-5-trifluoromethylpyridine-3-boronic acid

  • MDL number MFCD16660273
  • Cas Number : 1256345-53-5
  • Molecular Weight : 279.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2558

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2558
Availability details for
Quantity
  • Availability
  • Add to Cart