COM448613842 Combi-Blocks, Inc.

Oct-7-enylboronic acid

  • MDL number MFCD16660277
  • Cas Number : 1198772-66-5
  • Molecular Weight : 156.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2562

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2562
Availability details for
Quantity
  • Availability
  • Add to Cart