COM448613867 Combi-Blocks, Inc.

4-Bromo-2-chloro-5-methoxyphenylboronic acid

  • MDL number MFCD06659862
  • Cas Number : 850567-94-1
  • Molecular Weight : 265.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2596

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2596
Availability details for
Quantity
  • Availability
  • Add to Cart