COM448613869 Combi-Blocks, Inc.

2,4-Dichloro-5-methoxyphenylboronic acid

  • MDL number MFCD12546454
  • Cas Number : 431942-67-5
  • Molecular Weight : 220.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2598
Availability details for
Quantity
  • Availability
  • Add to Cart