COM448613883 Combi-Blocks, Inc.

2-Methyl-5-(N-morpholinylsulfonyl)phenylboronic acid

  • MDL number MFCD07363761
  • Cas Number : 871329-74-7
  • Molecular Weight : 285.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2613

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2613
Availability details for
Quantity
  • Availability
  • Add to Cart