COM448613888 Combi-Blocks, Inc.

N-Butyl 3-boronobenzenesulfonamide

  • MDL number MFCD07783858
  • Cas Number : 871329-78-1
  • Molecular Weight : 257.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2618

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2618
Availability details for
Quantity
  • Availability
  • Add to Cart