COM448613889 Combi-Blocks, Inc.

N-Cyclohexyl 3-boronobenzenesulfonamide

  • MDL number MFCD07783859
  • Cas Number : 871329-79-2
  • Molecular Weight : 283.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2619

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2619
Availability details for
Quantity
  • Availability
  • Add to Cart