COM448613890 Combi-Blocks, Inc.

N-Cyclopropyl 3-boronobenzenesulfonamide

  • MDL number MFCD08436035
  • Cas Number : 913835-28-6
  • Molecular Weight : 241.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2620

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2620
Availability details for
Quantity
  • Availability
  • Add to Cart