COM448613907 Combi-Blocks, Inc.

4-(3-Hydroxypropylcarbamoyl)phenylboronic acid

  • MDL number MFCD08235076
  • Cas Number : 913835-29-7
  • Molecular Weight : 223.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2637

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2637
Availability details for
Quantity
  • Availability
  • Add to Cart