COM448613953 Combi-Blocks, Inc.

3-(E-2-Nitrovinyl)phenylboronic acid

  • MDL number MFCD04115658
  • Cas Number : 850567-99-6
  • Molecular Weight : 193.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2683

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2683
Availability details for
Quantity
  • Availability
  • Add to Cart