COM448613967 Combi-Blocks, Inc.

3-(2-Ethoxy-2-oxoethoxy)phenylboronic acid

  • MDL number MFCD09878321
  • Cas Number : 957062-63-4
  • Molecular Weight : 224.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2703

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2703
Availability details for
Quantity
  • Availability
  • Add to Cart