COM448613978 Combi-Blocks, Inc.

2-Aminomethyl-4-fluorophenylboronic acid, HCl

  • MDL number MFCD04973742
  • Cas Number : 850568-02-4
  • Molecular Weight : 205.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2720

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2720
Availability details for
Quantity
  • Availability
  • Add to Cart