COM448614003 Combi-Blocks, Inc.

2-Carboxy-5-chlorophenylboronic acid

  • MDL number MFCD08235091
  • Cas Number : 874290-67-2
  • Molecular Weight : 200.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2747

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2747
Availability details for
Quantity
  • Availability
  • Add to Cart