COM448614014 Combi-Blocks, Inc.

3-Chloro-4-(cyclohexylcarbamoyl)phenylboronic acid

  • MDL number MFCD09475818
  • Cas Number : 957034-65-0
  • Molecular Weight : 281.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2763

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2763
Availability details for
Quantity
  • Availability
  • Add to Cart