COM448614038 Combi-Blocks, Inc.

4-(Isopropoxycarbonylamino)phenylboronic acid

  • MDL number MFCD17214243
  • Cas Number : 1033726-21-4
  • Molecular Weight : 223.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2791

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2791
Availability details for
Quantity
  • Availability
  • Add to Cart