COM448614039 Combi-Blocks, Inc.

2-Isopropoxy-5-trifluoromethylphenylboronic acid

  • MDL number MFCD07363786
  • Cas Number : 850593-12-3
  • Molecular Weight : 248.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2792

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2792
Availability details for
Quantity
  • Availability
  • Add to Cart