COM448614047 Combi-Blocks, Inc.

3-Fluoro-5-ethoxycarbonylphenylboronic acid

  • MDL number MFCD07363792
  • Cas Number : 871329-85-0
  • Molecular Weight : 211.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2800

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2800
Availability details for
Quantity
  • Availability
  • Add to Cart