COM448614059 Combi-Blocks, Inc.

4-Chloro-3-(t-butylcarbamoyl)phenylboronic acid

  • MDL number MFCD07783864
  • Cas Number : 871332-72-8
  • Molecular Weight : 255.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2816

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2816
Availability details for
Quantity
  • Availability
  • Add to Cart