COM448614070 Combi-Blocks, Inc.

3-Nitro-5-(pyrrolidine-1-carbonyl)phenylboronic acid

  • MDL number MFCD07783873
  • Cas Number : 871332-81-9
  • Molecular Weight : 264.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2828

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2828
Availability details for
Quantity
  • Availability
  • Add to Cart