COM448614074 Combi-Blocks, Inc.

3-(Methoxy(methyl)aminocarbonyl)-5-nitrophenylboronic acid

  • MDL number MFCD07783876
  • Cas Number : 871332-84-2
  • Molecular Weight : 254.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2832

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2832
Availability details for
Quantity
  • Availability
  • Add to Cart