COM448614075 Combi-Blocks, Inc.

3-(Cyclohexylaminocarbonyl)-5-nitrophenylboronic acid

  • MDL number MFCD07783877
  • Cas Number : 871332-85-3
  • Molecular Weight : 292.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2833

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2833
Availability details for
Quantity
  • Availability
  • Add to Cart