COM448614077 Combi-Blocks, Inc.

3-(t-Butylaminocarbonyl)-5-nitrophenylboronic acid

  • MDL number MFCD07783879
  • Cas Number : 871332-87-5
  • Molecular Weight : 266.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2835

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2835
Availability details for
Quantity
  • Availability
  • Add to Cart