COM448614079 Combi-Blocks, Inc.

3-(n-Butylaminocarbonyl)-5-nitrophenylboronic acid

  • MDL number MFCD07783881
  • Cas Number : 871332-89-7
  • Molecular Weight : 266.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2837

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2837
Availability details for
Quantity
  • Availability
  • Add to Cart