COM448614093 Combi-Blocks, Inc.

3-Fluoro-5-(propylcarbamoyl)phenylboronic acid

  • MDL number MFCD08235054
  • Cas Number : 874219-37-1
  • Molecular Weight : 225.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2855

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2855
Availability details for
Quantity
  • Availability
  • Add to Cart