COM448614095 Combi-Blocks, Inc.

3-(Cyclohexylcarbamoyl)-5-fluorophenylboronic acid

  • MDL number MFCD08235059
  • Cas Number : 874219-42-8
  • Molecular Weight : 265.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2857

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2857
Availability details for
Quantity
  • Availability
  • Add to Cart