COM448614119 Combi-Blocks, Inc.

3-(Benzyloxycarbonylamino)-4-fluorophenylboronic acid

  • MDL number MFCD19237159
  • Cas Number : 874219-58-6
  • Molecular Weight : 289.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2882

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2882
Availability details for
Quantity
  • Availability
  • Add to Cart