COM448614130 Combi-Blocks, Inc.

N-(3-Chloropropyl) 4-boronobenzenesulfonamide

  • MDL number MFCD08235065
  • Cas Number : 874219-48-4
  • Molecular Weight : 277.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2906

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2906
Availability details for
Quantity
  • Availability
  • Add to Cart