COM448614137 Combi-Blocks, Inc.

4-Methoxy-3-(N-morpholinylsulfonyl)phenylboronic acid

  • MDL number MFCD08056366
  • Cas Number : 871333-02-7
  • Molecular Weight : 301.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2917

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2917
Availability details for
Quantity
  • Availability
  • Add to Cart