COM448614139 Combi-Blocks, Inc.

4-Methoxy-3-(piperidin-1-ylsulfonyl)phenylboronic acid

  • MDL number MFCD08235037
  • Cas Number : 874219-18-8
  • Molecular Weight : 299.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2919

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2919
Availability details for
Quantity
  • Availability
  • Add to Cart