COM448614145 Combi-Blocks, Inc.

3-Isobutyramidophenylboronic acid

  • MDL number MFCD08235080
  • Cas Number : 874459-76-4
  • Molecular Weight : 207.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2936

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2936
Availability details for
Quantity
  • Availability
  • Add to Cart