COM448614147 Combi-Blocks, Inc.

2-Formyl-4,5-dimethoxyphenylboronic acid

  • MDL number MFCD16660278
  • Cas Number : 1005346-96-2
  • Molecular Weight : 210.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2941

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2941
Availability details for
Quantity
  • Availability
  • Add to Cart