COM448614150 Combi-Blocks, Inc.

2-(N,N-Diethylaminocarbonyl)phenylboronic acid

  • MDL number MFCD01318988
  • Cas Number : 129112-21-6
  • Molecular Weight : 221.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2951

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2951
Availability details for
Quantity
  • Availability
  • Add to Cart