COM448614151 Combi-Blocks, Inc.

2-(Morpholine-4-carbonyl)phenylboronic acid

  • MDL number MFCD08235036
  • Cas Number : 874219-17-7
  • Molecular Weight : 235.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2954

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-2954
Availability details for
Quantity
  • Availability
  • Add to Cart