COM448614169 Combi-Blocks, Inc.

4-(2-Cyanoethylaminocarbonyl)phenylboronic acid

  • MDL number MFCD04115669
  • Cas Number : 850568-16-0
  • Molecular Weight : 218.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3016

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3016
Availability details for
Quantity
  • Availability
  • Add to Cart