COM448614173 Combi-Blocks, Inc.

Ethyl 3-(4-boronobenzamido)propionate

  • MDL number MFCD04115673
  • Cas Number : 850568-19-3
  • Molecular Weight : 265.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3020

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3020
Availability details for
Quantity
  • Availability
  • Add to Cart