COM448614175 Combi-Blocks, Inc.

4-(4-Methylpiperidine-1-carbonyl)phenylboronic acid

  • MDL number MFCD04115676
  • Cas Number : 850568-21-7
  • Molecular Weight : 247.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3023

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3023
Availability details for
Quantity
  • Availability
  • Add to Cart