COM448614188 Combi-Blocks, Inc.

N-(Thiazoline-2-yl) 4-boronobenzamide

  • MDL number MFCD06659881
  • Cas Number : 850568-30-8
  • Molecular Weight : 250.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3044

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3044
Availability details for
Quantity
  • Availability
  • Add to Cart