COM448614193 Combi-Blocks, Inc.

4-(Benzyl(methyl)carbamoyl)phenylboronic acid

  • MDL number MFCD08235067
  • Cas Number : 874219-49-5
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3049

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3049
Availability details for
Quantity
  • Availability
  • Add to Cart