COM448614201 Combi-Blocks, Inc.

3-(N-Ethylaminocarbonyl)phenylboronic acid

  • MDL number MFCD04115689
  • Cas Number : 850567-21-4
  • Molecular Weight : 193.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3057

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3057
Availability details for
Quantity
  • Availability
  • Add to Cart