COM448614208 Combi-Blocks, Inc.

3-(Cyclopentylaminocarbonyl)phenylboronic acid

  • MDL number MFCD04115695
  • Cas Number : 850567-24-7
  • Molecular Weight : 233.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3064

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3064
Availability details for
Quantity
  • Availability
  • Add to Cart