COM448614215 Combi-Blocks, Inc.

3-Allylaminocarbonylphenylboronic acid

  • MDL number MFCD04115702
  • Cas Number : 850567-29-2
  • Molecular Weight : 205.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3071

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3071
Availability details for
Quantity
  • Availability
  • Add to Cart