COM448614222 Combi-Blocks, Inc.

N-(1H,1,2,4-Triazol-3-yl) 3-boronobenzamide

  • MDL number MFCD07363798
  • Cas Number : 871333-05-0
  • Molecular Weight : 232.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3085

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3085
Availability details for
Quantity
  • Availability
  • Add to Cart