COM448614224 Combi-Blocks, Inc.

3-(4-Cyanophenyl)aminocarbonylphenylboronic acid

  • MDL number MFCD06659886
  • Cas Number : 850567-36-1
  • Molecular Weight : 266.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3090

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3090
Availability details for
Quantity
  • Availability
  • Add to Cart