COM448614225 Combi-Blocks, Inc.

3-(Thiomorpholine-4-carbonyl)phenylboronic acid

  • MDL number MFCD06659887
  • Cas Number : 850567-37-2
  • Molecular Weight : 251.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3091

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3091
Availability details for
Quantity
  • Availability
  • Add to Cart