COM448614227 Combi-Blocks, Inc.

3-(1H-Tetrazol-5-yl-carbamoyl)phenylboronic acid, HCl

  • MDL number MFCD06659888
  • Cas Number : 850567-38-3
  • Molecular Weight : 269.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3095

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3095
Availability details for
Quantity
  • Availability
  • Add to Cart